Learn More
Enterprise Technology Solutions Simple Green Multipurpose Cleaning Towels,10"x11-3/4, 75 Wipes
SDP
Catalog No. 5000789967
Encompass_Preferred
Safety Towels are larger-sized wipes for heavy-duty, multipurpose cleaning. They are pre-moistened with Crystal Simple Green, a powerful cleaner and degreaser that is nontoxic, nonabrasive and noncorrosive. The durable, dual-sided wipes feature a scouring side for tough scrubbing and a smooth side for a nonabrasive, lint-free shine. The wipes feature excellent wicking and fluid-holding capacity for cleaning liquid spills and are thermally bonded and melt-blown to ensure durability and resistance to rot and mildew. Easily remove grease, oil, food spills, wet paint, carbon, graphite, tar, soot, grime, most inks and more. They are safe for use on all washable surfaces including metals, concrete, rubber, finished wood, tile, laminates, concrete, stone, porcelain, carpets and other industrial surfaces. The cleaning solution contains no added color or scent and is NSF-registered for use in food-processing facilities and commercial kitchens.
Catalog No. 50-007-89967 Supplier Enterprise Technology Solutions Supplier No. SMP13351
Safety Towels are larger-sized wipes for heavy-duty, multipurpose cleaning. They are pre-moistened with Crystal Simple Green, a powerful cleaner and degreaser that is nontoxic, nonabrasive and noncorrosive. The durable, dual-sided wipes feature a scouring side for tough scrubbing and a smooth side for a nonabrasive, lint-free shine. The wipes feature excellent wicking and fluid-holding capacity for cleaning liquid spills and are thermally bonded and melt-blown to ensure durability and resistance to rot and mildew. Easily remove grease, oil, food spills, wet paint, carbon, graphite, tar, soot, grime, most inks and more. They are safe for use on all washable surfaces including metals, concrete, rubber, finished wood, tile, laminates, concrete, stone, porcelain, carpets and other industrial surfaces. The cleaning solution contains no added color or scent and is NSF-registered for use in food-processing facilities and commercial kitchens.