Learn More
Enterprise Technology Solutions Simple Green Pro HD Heavy-Duty Cleaner, 1Gal, 2/CT, Purple
SDP
Catalog No. 5000830143
Encompass_Preferred
Heavy-duty cleaner features a noncorrosive, professional-grade formula that breaks down tough grease, grime and oily stains for multipurpose home and do-it-yourself applications. Versatile design can be used manually or in equipment, including parts washers, dip tanks, pressure washers and ultrasonics. The cost-effective concentrate can be custom diluted for specific applications and creates up to 14 gallons of cleaner. Heavy-duty cleaner is perfect for floors, painted surfaces, metal surfaces, engines, wheels, tools, heavy equipment, grease/oil stains, vehicles, ovens, grills and more.
Catalog No. 50-008-30143 Supplier Enterprise Technology Solutions Supplier No. SMP213421
Heavy-duty cleaner features a noncorrosive, professional-grade formula that breaks down tough grease, grime and oily stains for multipurpose home and do-it-yourself applications. Versatile design can be used manually or in equipment, including parts washers, dip tanks, pressure washers and ultrasonics. The cost-effective concentrate can be custom diluted for specific applications and creates up to 14 gallons of cleaner. Heavy-duty cleaner is perfect for floors, painted surfaces, metal surfaces, engines, wheels, tools, heavy equipment, grease/oil stains, vehicles, ovens, grills and more.