Learn More
Simple Nutrients in Seawater, Whole Volume, Certified Reference Material, MilliporeSigma™ Supelco™
Catalog No. 111004801 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
500 mL
Catalog No. Quantity
11-100-4801 500 mL
1 options
Catalog No. 11-100-4801 Supplier MilliporeSigma Supelco Supplier No. QC3179500ML
Add to Cart
Add to Cart
This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.
TRUSTED_SUSTAINABILITY
Specifications
Quantity 500 mL
Grade Certified Reference Material
Concentration or Composition (by Analyte or Components) Ammonia as N 0.65-19 mg/L; Nitrate as N 0.25-40 mg/L; Nitrate+nitrite as N 0.25-40 mg/L; Nitrite as N 0.4-4 mg/L; Orthophosphate as P 0.5-5.5 mg/L