Learn More
Gold Biotechnology Inc Simple Stop™ 1 Phosphatase Inhibitor Cocktail 1 mL
SDP
Supplier:  Gold Biotechnology Inc GB4501
Encompass
We have reengineered our GoldBio Simple Stop™ 1 to have significantly better protection against Acid Phosphatases as well as enhanced protection against Protein Phosphatase.
Catalog No. NC2596282