Learn More
Abcam SimpleStep ELISA Custom ELISA Kit, 5 x 96 Tests
Supplier:  Abcam AB2705525X96TESTS
Encompass_Preferred
SimpleStep ELISA Custom ELISA Kit allows you to develop your own SimpleSte ELISA using any antibody pair. - No plate coating needed - Possibility to measure different target on the same plate - All components included, just add the antibody pair - Streamline your ELISA to a single-wash format if desired
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC2111531