Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Simplicon™ Human iPS Cell Line

Catalog No. SCC271MI Shop All MilliporeSigma Products
Change view
Click to view available options
No. of Cells:
≥ 1 x 106 Cells/Vial
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Cells
SCC271MI ≥ 1 x 106 Cells/Vial
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SCC271MI Supplier MilliporeSigma™ Supplier No. SCC271
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Highly characterized integration-free human induced pluripotent stem cell (iPSC) line

  • Generated using Simplicon™ RNA reprogramming technology
  • Human induced pluripotent stem cells (iPSCs), have the capacity to give rise to differentiated progeny representative of all three germ layers (ectoderm, endoderm, and mesoderm)

Contains genetically modified organisms (GMO)

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Stored in liquid nitrogen, Can be cultured for 10 passages after initial thawing without significantly affecting cell marker expression and functionality
Cell Type Induced Pluripotent Stem Cells (iPSC)
For Use With (Application) Cell Culture, Stem Cell Culture
No. of Cells ≥ 1 x 106 Cells/Vial

For research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.