Learn More
Azer Scientific Simply Pure™ Glucose Tolerance Beverages
SDP
Catalog No. 22026371
Change view
Click to view available options
Quantity:
50 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-026-371 100 g
22-026-362 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 22-026-371 Supplier Azer Scientific Supplier No. 10SP100
Add to Cart
Add to Cart
This item is not returnable. View return policy
A reliable alternative for patients with an allergy to citric acid or other additives in flavored glucose drinks
  • Beverages provide a premeasured glucose load of either 50, 75 or 100 grams
  • Beverage contains no flavor, color, citric acid, dairy, or gluten
  • Certified Kosher Product
  • Manufactured according to WHO and ADA standards
Beverage should be kept out of direct sunlight and high heat and humidity areas. Excessive exposure to high heat and humidity or direct sunlight may affect the taste and product stability.
Specifications
Certifications/Compliance FDA, WHO, ADA
Content And Storage Room Temperature
Quantity 100 g