Learn More
Princeton Cryotech T314-2100 Cryostore Box - 4 Pack, Red
Catalog No. NC1391287
Encompass_Preferred
100 cell box fits 1 to 2 ml vials. Made of extra strong polycarbonate, these durable cryogenic storage boxes are designed to be used at temperatures between -196 °C and +121 °C and are autoclavable at 120 °C, psig (1 bar) for 20 minutes. A transparent cover allows the user to see the contents of the box , and is keyed to the base to prevent misalignment. Printed with a series of squares numbered from 1 - 100, the surface accepts writing with markers, facilitating inventory control. Removal of vials facilitated by an innovative vial picker supplied with each storage box.
Catalog No. NC1391287 Supplier Princeton Cryotech Supplier No. T3142100G
100 cell box fits 1 to 2 ml vials. Made of extra strong polycarbonate, these durable cryogenic storage boxes are designed to be used at temperatures between -196 °C and +121 °C and are autoclavable at 120 °C, psig (1 bar) for 20 minutes. A transparent cover allows the user to see the contents of the box , and is keyed to the base to prevent misalignment. Printed with a series of squares numbered from 1 - 100, the surface accepts writing with markers, facilitating inventory control. Removal of vials facilitated by an innovative vial picker supplied with each storage box.