Learn More
Sklar Instruments Sims Uterine Sound, Sklar, 12-1/2 in. Sims Uterine Sound CM Graduation Silver Plated. EA/1.
SDP
Catalog No. 501436807 Shop All Sklar Instruments Products
Encompass_Preferred
Sims Uterine Sound, Sklar, 12-1/2 in. Sims Uterine Sound CM Graduation Silver Plated. Sklar's Sims Uterine Sound is a malleable, silver plated OB/GYN instrument used for probing and dilating the uterus as well as measuring the length of the cervical canal. It is very similar to the Simpson uterine sound. The instrument is available with or without graduated centimeter markings. This product is straight, but malleable and can be curved as necessary. The handle is a short, smooth thumb grip that features 3 vertical grooves for improved grip. It also has graduated centimeter markings. Overall, the length is 12-1/2 inches.
This item is not returnable. View return policy
Catalog No. 50-143-6807 Supplier Sklar Instruments Supplier No. 905314
Sims Uterine Sound, Sklar, 12-1/2 in. Sims Uterine Sound CM Graduation Silver Plated. Sklar's Sims Uterine Sound is a malleable, silver plated OB/GYN instrument used for probing and dilating the uterus as well as measuring the length of the cervical canal. It is very similar to the Simpson uterine sound. The instrument is available with or without graduated centimeter markings. This product is straight, but malleable and can be curved as necessary. The handle is a short, smooth thumb grip that features 3 vertical grooves for improved grip. It also has graduated centimeter markings. Overall, the length is 12-1/2 inches.