Learn More
3B Scientific™ SimShirt System Auscultation Simulator
GREENER_CHOICESDPSCI_ED
Catalog No. S58111 Shop All 3B Scientific Products
Change view
Click to view available options
Size:
Medium
Large
X-Large
2 X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
S58111 Medium
S58108 Large
S58109 X-Large
S58110 2 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. S58111 Supplier 3B Scientific™ Supplier No. 1021552
Add to Cart
Add to Cart
Request a Quote
This item is not returnable. View return policy
When worn by the standardized patient, the SimShirt simulates physiological conditions to test diagnostic and procedural skills. It is controlled wirelessly with easy to use software and responds in real time to diagnosis and treatment with direct feedback.
  • The Shirt offers a basic version with all features of the Bionic Hybrid Simulator, except EKG and blood pressure.
  • Includes SimScope Wifi and tablet. Reusable and washable (30°C), available in different sizes
  • Can be worn by students and manikins
  • Simulation of heart, breath, and bowel sounds
  • RFID sensor tags integrated
  • Simulates multiple physiological conditions
  • Real time diagnostics and treatment capabilities
Specifications
Product Type Patient Examination Simulator
For Use With (Application) Train and Grow Confidence in Patient Examination
Size Medium