Learn More
Selleck Chemical LLC Simufilam dihydrochloride
SDP
Catalog No. 503136007
Encompass_Preferred
Simufilam (PTI-125) Dihydrochloride is a small molecule modulator that preferentially binds altered FLNA and restores its native conformation restoring receptor and synaptic activities and reducing its 7 nAChR/TLR4 associations and downstream pathologies
Catalog No. 50-313-6007 Supplier Selleck Chemical LLC Supplier No. E007625MG
Simufilam (PTI-125) Dihydrochloride is a small molecule modulator that preferentially binds altered FLNA and restores its native conformation restoring receptor and synaptic activities and reducing its 7 nAChR/TLR4 associations and downstream pathologies