Learn More
Medchemexpress LLC Simufilam hydrochloride | 2480226-07-9 | 99.9% | 295.81 | 10 MG
SDP
Catalog No. 5000365541
Encompass_Preferred
Simufilam hydrochloride (PTI-125 hydrochloride) is an orally active FLNA modulator. It restores NMDAR signaling and Arc expression, inhibits overactive mTOR signaling by restoring the normal conformation of FLNA, improves insulin sensitivity, and reduces Aβ42-induced neuroinflammation and tau protein hyperphosphorylation. It can be used for research of Alzheimer's disease.
  • Acts as an orally active FLNA modulator
  • Restores NMDAR signaling and Arc expression
  • Inhibits overactive mTOR signaling by restoring normal FLNA conformation
  • Improves insulin sensitivity
  • Reduces Aβ42-induced neuroinflammation and tau protein hyperphosphorylation
  • Useful for Alzheimer's disease research
Catalog No. 50-003-65541 Supplier Medchemexpress LLC Supplier No. 10MGHY139142B
Simufilam hydrochloride (PTI-125 hydrochloride) is an orally active FLNA modulator. It restores NMDAR signaling and Arc expression, inhibits overactive mTOR signaling by restoring the normal conformation of FLNA, improves insulin sensitivity, and reduces Aβ42-induced neuroinflammation and tau protein hyperphosphorylation. It can be used for research of Alzheimer's disease.
  • Acts as an orally active FLNA modulator
  • Restores NMDAR signaling and Arc expression
  • Inhibits overactive mTOR signaling by restoring normal FLNA conformation
  • Improves insulin sensitivity
  • Reduces Aβ42-induced neuroinflammation and tau protein hyperphosphorylation
  • Useful for Alzheimer's disease research