Learn More
Medchemexpress LLC Simufilam hydrochloride (PTI-125 hydrochloride) | 2480226-07-9 | 99.9% | 295.81 | 25 MG
SDP
Catalog No. 5000350646
Encompass_Preferred
Simufilam hydrochloride (PTI-125 hydrochloride) is an orally active FLNA modulator. It restores NMDAR signaling and Arc expression. It inhibits overactive mTOR signaling by restoring the normal conformation of FLNA, improves insulin sensitivity, reduces Aβ42-induced neuroinflammation, and tau protein hyperphosphorylation. This product can be used for research related to Alzheimer's disease.
  • Reduces FLNA-α7nAChR/TLR4 associations and dissociates Aβ42-α7nAChR complexes.
  • Restores α7nAChR/NMDAR/IR function and NMDAR signaling and arc expression in human AD brain slices.
  • Decreases tau phosphorylation, Aβ42 aggregates, and inflammatory cytokines in mouse models.
  • Improves NMDAR/IR function, synaptic plasticity, spatial/working memory, and nesting behavior in 3xTg AD mice.
Catalog No. 50-003-50646 Supplier Medchemexpress LLC Supplier No. 25MGHY139142B
Simufilam hydrochloride (PTI-125 hydrochloride) is an orally active FLNA modulator. It restores NMDAR signaling and Arc expression. It inhibits overactive mTOR signaling by restoring the normal conformation of FLNA, improves insulin sensitivity, reduces Aβ42-induced neuroinflammation, and tau protein hyperphosphorylation. This product can be used for research related to Alzheimer's disease.
  • Reduces FLNA-α7nAChR/TLR4 associations and dissociates Aβ42-α7nAChR complexes.
  • Restores α7nAChR/NMDAR/IR function and NMDAR signaling and arc expression in human AD brain slices.
  • Decreases tau phosphorylation, Aβ42 aggregates, and inflammatory cytokines in mouse models.
  • Improves NMDAR/IR function, synaptic plasticity, spatial/working memory, and nesting behavior in 3xTg AD mice.