Learn More
Medchemexpress LLC Simufilam (PTI-125) | 1224591-33-6 | 99.8% | 259.35 | 1 ML
SDP
Catalog No. 5000347824
Encompass_Preferred
Simufilam (PTI-125) is an orally active FLNA modulator. It restores NMDAR signaling and Arc expression, inhibits overactive mTOR signaling by restoring the normal conformation of FLNA, improves insulin sensitivity, and reduces Aβ42-induced neuroinflammation and tau protein hyperphosphorylation. This modulator can be used for research of Alzheimer's disease.
  • Restores NMDAR signaling and Arc expression
  • Inhibits overactive mTOR signaling
  • Improves insulin sensitivity
  • Reduces Aβ42-induced neuroinflammation
  • Reduces tau protein hyperphosphorylation
Catalog No. 50-003-47824 Supplier Medchemexpress LLC Supplier No. HY13914210MM/1ML
Simufilam (PTI-125) is an orally active FLNA modulator. It restores NMDAR signaling and Arc expression, inhibits overactive mTOR signaling by restoring the normal conformation of FLNA, improves insulin sensitivity, and reduces Aβ42-induced neuroinflammation and tau protein hyperphosphorylation. This modulator can be used for research of Alzheimer's disease.
  • Restores NMDAR signaling and Arc expression
  • Inhibits overactive mTOR signaling
  • Improves insulin sensitivity
  • Reduces Aβ42-induced neuroinflammation
  • Reduces tau protein hyperphosphorylation