Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ Simulated Blood Spatter
GREENER_CHOICESCI_ED

Catalog No. S29560 Shop All Aldon Products
Change view
Click to view available options
Class Size:
15 Lab Groups
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S29560 15 Lab Groups
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S29560 Supplier Innovating Science™ Supplier No. IS9075
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Innovating Science™ Simulated Blood Spatter is not corn syrup based and will dry on material allowing students to utilize the dried blood on objects for exams or other lab measurements.

  • Useful for various types of blood spatter analysis including: the effect of height on blood droplets, the appearance of blood that impacts a surface at varying angles, some common blood spatter patterns that may be present at a crime scene, the effect of impact velocity on blood spatter, and the manner in which various surface types/textures may affect the appearance of blood evidence
  • For complete Blood Spatter Kit, see S12833

Specifications

Color White
Grade 7 to 14
Age 13 to 20
For Use With (Application) Forensics class (for groups of 15)
Includes Teachers Manual and Student Guide
Quantity 1 L
Dimensions (L x W x H) 3.25 x 3.25 x 9 in.
Class Size 15 Lab Groups
Product Type Simulated Blood Spatter
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.