Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Simulated Intestinal Fluid (without Pancreatin), USP XXII Formulation, Ricca Chemical
SDP

Catalog No. 71097516 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
20 L
Packaging:
Cubitainer
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
710975-16 500 mL Natural Poly Bottle
R710975020F 20 L Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 710975-16 Supplier Ricca Chemical Company Supplier No. 7109.7516
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Simulated Intestinal Fluid (without Pancreatin)

Simulated Intestinal Fluid (without Pancreatin), CAS Number-7778-77-0, 1310-73-2, 500mL, 100.0, 0.66, 0.29, 97.0, 0.64, 0.27, Colorless, USP XXII Formulation, Poly natural, Liquid

Specifications

Chemical Name or Material Simulated Intestinal Fluid (without Pancreatin)
Color Colorless
Name Note USP XXII Formulation
CAS 7732-18-5, 7778-77-0, 1310-73-2
CAS Min % 97.0, 0.64, 0.27
CAS Max % 100.0, 0.66, 0.29
Grade Laboratory
Packaging Natural Poly Bottle
pH 7.4 to 7.6
Quantity 500 mL
Physical Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.