Learn More
Simulated Intestinal Fluid (without Pancreatin), USP XXII Formulation, Ricca Chemical
Shop All Ricca Chemical Company Products

Description
Simulated Intestinal Fluid (without Pancreatin), CAS Number-7778-77-0, 1310-73-2, 500mL, 100.0, 0.66, 0.29, 97.0, 0.64, 0.27, Colorless, USP XXII Formulation, Poly natural, Liquid
Specifications
Specifications
| Chemical Name or Material | Simulated Intestinal Fluid USP XXII |
| Color | Colorless Liquid |
| CAS | 7732-18-5, 7778-77-0, 1310-73-2 |
| CAS Max % | 99.17, 0.68, 0.15 |
| Grade | Laboratory |
| Packaging | Cubitainer |
| pH | 7.5 |
| Quantity | 20 L |
| Sterility | Non-sterile |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.