Learn More
Medchemexpress LLC HY-100888 2mg Medchemexpress, Simurosertib CAS:1330782-76-7 Purity:>98%
SDP
Catalog No. 502028639
Encompass_Preferred
Medchemexpress, HY-100888 2mg Simurosertib CAS:1330782-76-7 Simurosertib (TAK-931) is an orally active, selective and ATP-competitive cell division cycle 7 (CDC7) kinase inhibitor, with an IC50 of <0.3 nM. Simurosertib has anti-cancer activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-8639 Supplier Medchemexpress LLC Supplier No. HY1008882MG
Medchemexpress, HY-100888 2mg Simurosertib CAS:1330782-76-7 Simurosertib (TAK-931) is an orally active, selective and ATP-competitive cell division cycle 7 (CDC7) kinase inhibitor, with an IC50 of <0.3 nM. Simurosertib has anti-cancer activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.