Learn More
Medchemexpress LLC HY-100888 50mg Medchemexpress, Simurosertib CAS:1330782-76-7 Purity:>98%
SDP
Catalog No. 501931988
Encompass_Preferred
Medchemexpress, HY-100888 50mg Simurosertib CAS:1330782-76-7 Simurosertib (TAK-931) is a selective cycle 7 (CDC7) kinase inhibitor, with an IC 50 <0.3 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-1988 Supplier Medchemexpress LLC Supplier No. HY10088850MG
Medchemexpress, HY-100888 50mg Simurosertib CAS:1330782-76-7 Simurosertib (TAK-931) is a selective cycle 7 (CDC7) kinase inhibitor, with an IC 50 <0.3 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.