Learn More
Medchemexpress LLC HY-17502 100mg , Simvastatin CAS:79902-63-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY17502100MG
Encompass_Preferred
HY-17502 100mg Simvastatin CAS: 79902-63-9 Simvastatin (MK 733) is a competitive inhibitor of HMG-CoA reductase with a Ki of 0.2 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9774