Learn More
Selleck Chemical LLC Simvastatin 25mg 79902-63-9 MK-0733
SDP
Supplier:  Selleck Chemical LLC S179225MG
Encompass_Preferred
Simvastatin (MK-0733) is a competitive inhibitor of HMG-CoA reductase with Ki of 0.1-0.2 nM in cell-free assays. Simvastatin induces ferroptosis, mitophagy, autophagy and apoptosis. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-1641