Learn More
Medchemexpress LLC HY-17502 500mg , Simvastatin MK 733 CAS:79902-63-9 Purity:98%
SDP
Catalog No. 501492363
Encompass_Preferred
Medchemexpress, HY-17502 500mg Simvastatin MK 733 CAS:79902-63-9 Formula:C25H38O5 Ki: 0.2 nM (HMG-CoA reductase) Purity:98% Simvastatin is a competitive inhibitor of HMG-CoA reductase with a K i of 0.2 nM. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-2363 Supplier Medchemexpress LLC Supplier No. HY17502500MG
Medchemexpress, HY-17502 500mg Simvastatin MK 733 CAS:79902-63-9 Formula:C25H38O5 Ki: 0.2 nM (HMG-CoA reductase) Purity:98% Simvastatin is a competitive inhibitor of HMG-CoA reductase with a K i of 0.2 nM. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.