Learn More
Cayman Chemical Simvastatin, 79902-63-9, 5 mg
SDP
Supplier:  Cayman Chemical 100103445
Encompass_Preferred
Molecular Formula C25H38O5, Purity ≥98%, Formula Weight 418.6, Synonyms: MK-733; A potent, competitive inhibitor of HMG-CoA reductase (Ki = 0.12 nM); 8 mg/kg/day for 18 days reduces plasma cholesterol levels by 33% in dogs; suppresses TNF-induced NF-B activation (IC50 ~ 13 M).
Catalog No. NC1512722
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.