Learn More
AdipoGen Simvastatin (50 mg)
SDP
Supplier:  AdipoGen AGCN20052M050
Encompass_Preferred
Chemical. CAS: 79902-63-9. Formula: C25H38O5. MW: 418.6. Synthetic. Synthetic analog of Lovastatin (Prod. No. AG-CN2-0051 http://www.adipogen.com/ag-cn2-0051/lovastatin.html ). Potent competitive HMG-CoA reductase inhibitor. Anti-hypercholesterolemic agent. Cholesterol/isoprenoid biosynthesis inhibitor. Blocks the production of mevalonate, a critical compound in the production of cholesterol and isoprenoids. Inhibits the isoprenylation of Ras and Rho family GTPases. Smooth muscle cell proliferation inhibitor. Anti-adhesive, immunomodulatory and anti-inflammatory compound. Proteasome modulator. Stimulates bone formation. Apoptosis inducer. Anticancer compound. Increases cellular resistance to anticancer agents such as doxorubicin and etoposides (Prod. No. AG-CR1-3572 http://www.adipogen.com/ag-cr1-3572/etoposide.html ). Suppresses ICAM-1-LFA-1 interactions, which blocks virus replication and infection. Anti-hypertensive agent. Suppresses TNF-induced NF-kappaB activation.
Catalog No. 50-205-3395