Learn More
Medchemexpress LLC Simvastatin | 79902-63-9 | 00072007 | 99.0% | 418.57 g/mol | C25H38O5 | 10 MG
SDP
Catalog No. 5000298705
Encompass_Preferred
Simvastatin (Standard) is an analytical reference standard of simvastatin supplied for research and analytical applications. It serves as a calibration and validation material for quantitative assays and method development; simvastatin is a competitive inhibitor of HMG-CoA reductase (Ki ≈ 0.2 nM). This material is intended for laboratory use only and not for human consumption.
  • High purity (~99.0%) for accurate analytical results.
  • Suitable for method development and calibration.
  • Provided in small and larger package sizes for flexibility.
  • Known molecular weight and formula for reference calculations.
  • Intended for research use and analytical applications only.
Catalog No. 50-002-98705 Supplier Medchemexpress LLC Supplier No. HY17502R10MG
Simvastatin (Standard) is an analytical reference standard of simvastatin supplied for research and analytical applications. It serves as a calibration and validation material for quantitative assays and method development; simvastatin is a competitive inhibitor of HMG-CoA reductase (Ki ≈ 0.2 nM). This material is intended for laboratory use only and not for human consumption.
  • High purity (~99.0%) for accurate analytical results.
  • Suitable for method development and calibration.
  • Provided in small and larger package sizes for flexibility.
  • Known molecular weight and formula for reference calculations.
  • Intended for research use and analytical applications only.