Learn More
Medchemexpress LLC Simvastatin acid | 121009-77-6 | 95.5% | 25 MG
SDP
Catalog No. 5000234638
Encompass_Preferred
Simvastatin acid is the hydrolyzed, active form of simvastatin and a potent HMG-CoA reductase inhibitor used in biochemical and cellular research. It has reported effects on reactive oxygen species and OATP3A1 expression in cardiomyocyte models.
  • Active hydrolyzed form of simvastatin
  • Inhibits HMG-CoA reductase (HMGCR)
  • Reduces indoxyl sulfate-mediated reactive oxygen species in cardiomyocytes
  • Available as a 25 mg research-grade solid
  • Includes analytical documentation such as COA, HNMR, LCMS, and RP-HPLC
Catalog No. 50-002-34638 Supplier Medchemexpress LLC Supplier No. HY11969525MG
Simvastatin acid is the hydrolyzed, active form of simvastatin and a potent HMG-CoA reductase inhibitor used in biochemical and cellular research. It has reported effects on reactive oxygen species and OATP3A1 expression in cardiomyocyte models.
  • Active hydrolyzed form of simvastatin
  • Inhibits HMG-CoA reductase (HMGCR)
  • Reduces indoxyl sulfate-mediated reactive oxygen species in cardiomyocytes
  • Available as a 25 mg research-grade solid
  • Includes analytical documentation such as COA, HNMR, LCMS, and RP-HPLC