Learn More
Medchemexpress LLC Simvastatin acid | 121009-77-6 | 95.5% | 50 MG
SDP
Catalog No. 5000234579
Encompass_Preferred
Simvastatin acid is the hydrolyzed acid form of simvastatin that acts as an HMG-CoA reductase inhibitor and is supplied for biochemical and cell-based research applications; not for human use.
  • Hydrolyzed active acid form of simvastatin.
  • Inhibits HMG-CoA reductase activity in biochemical assays.
  • Reduces indoxyl sulfate-mediated reactive oxygen species in cardiomyocytes.
  • Molecular formula C25H40O6, molecular weight 436.58 g/mol.
  • Available in milligram quantities (1 mg-100 mg) for research applications.
  • Typical purity around 95.5% (supplier listed).
Catalog No. 50-002-34579 Supplier Medchemexpress LLC Supplier No. HY11969550MG
Add to Cart
Add to Cart
Simvastatin acid is the hydrolyzed acid form of simvastatin that acts as an HMG-CoA reductase inhibitor and is supplied for biochemical and cell-based research applications; not for human use.
  • Hydrolyzed active acid form of simvastatin.
  • Inhibits HMG-CoA reductase activity in biochemical assays.
  • Reduces indoxyl sulfate-mediated reactive oxygen species in cardiomyocytes.
  • Molecular formula C25H40O6, molecular weight 436.58 g/mol.
  • Available in milligram quantities (1 mg-100 mg) for research applications.
  • Typical purity around 95.5% (supplier listed).