Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Simvastatin acid ammonium | 139893-43-9 | 99.7% | 1 ML
SDP

Catalog No. 5000233093
Encompass_Preferred

Simvastatin acid ammonium is the hydroxy-acid form of simvastatin supplied as the ammonium salt. It is a potent HMG-CoA reductase inhibitor used in research to study cholesterol metabolism and oxidative stress. The compound is offered as a ready-to-use 10 mM solution in DMSO (1 mL) and in multiple solid pack sizes.

  • Potent HMG-CoA reductase inhibitor active in cellular assays.
  • Supplied as a 10 mM solution in DMSO, 1 mL ready to use.
  • High purity: 99.7%.
  • Molecular weight 453.61 g/mol; formula C25H43NO6.
  • Reconstitution guidance: warming and ultrasonic treatment improves solubility in DMSO; use newly opened DMSO.

Catalog No. 50-002-33093 Supplier Medchemexpress LLC Supplier No. HY119695A10MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Simvastatin acid ammonium is the hydroxy-acid form of simvastatin supplied as the ammonium salt. It is a potent HMG-CoA reductase inhibitor used in research to study cholesterol metabolism and oxidative stress. The compound is offered as a ready-to-use 10 mM solution in DMSO (1 mL) and in multiple solid pack sizes.

  • Potent HMG-CoA reductase inhibitor active in cellular assays.
  • Supplied as a 10 mM solution in DMSO, 1 mL ready to use.
  • High purity: 99.7%.
  • Molecular weight 453.61 g/mol; formula C25H43NO6.
  • Reconstitution guidance: warming and ultrasonic treatment improves solubility in DMSO; use newly opened DMSO.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.