Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Simvastatin acid ammonium | 139893-43-9 | 99.7% | 10 MG
SDP

Catalog No. 5000234253
Encompass_Preferred

Simvastatin acid ammonium is the ammonium salt of simvastatin acid and acts as an HMG-CoA reductase inhibitor used in biochemical and pharmacological research to study cholesterol biosynthesis and related cellular pathways. It is provided as a purified solid suitable for in vitro studies.

  • High purity 99.7%
  • Molecular formula C25H43NO6 and molecular weight 453.61 g/mol
  • Soluble in DMSO
  • Supplied as a small, research-grade quantity suitable for in vitro assays
  • Used to study HMG-CoA reductase activity and cholesterol metabolism

Catalog No. 50-002-34253 Supplier Medchemexpress LLC Supplier No. HY119695A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Simvastatin acid ammonium is the ammonium salt of simvastatin acid and acts as an HMG-CoA reductase inhibitor used in biochemical and pharmacological research to study cholesterol biosynthesis and related cellular pathways. It is provided as a purified solid suitable for in vitro studies.

  • High purity 99.7%
  • Molecular formula C25H43NO6 and molecular weight 453.61 g/mol
  • Soluble in DMSO
  • Supplied as a small, research-grade quantity suitable for in vitro assays
  • Used to study HMG-CoA reductase activity and cholesterol metabolism

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.