Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Simvastatin acid ammonium | 139893-43-9 | 99.7% | 25 MG
SDP

Catalog No. 5000234053
Encompass_Preferred

Simvastatin acid ammonium is the ammonium salt of simvastatin hydroxy acid, the active hydrolyzed form of simvastatin. It is a potent HMG-CoA reductase inhibitor supplied as a research-grade reference compound with high purity for biochemical and analytical studies. For research use only.

  • Potent HMG-CoA reductase inhibitor.
  • High purity suitable for analytical and biochemical studies.
  • Available as a 25 mg pack for small-scale assays.
  • Molecular weight 453.61 g·mol⁻1 and formula C25H43NO6.
  • Used to study cholesterol biosynthesis, ROS modulation, and transporter expression.

Catalog No. 50-002-34053 Supplier Medchemexpress LLC Supplier No. HY119695A25MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Simvastatin acid ammonium is the ammonium salt of simvastatin hydroxy acid, the active hydrolyzed form of simvastatin. It is a potent HMG-CoA reductase inhibitor supplied as a research-grade reference compound with high purity for biochemical and analytical studies. For research use only.

  • Potent HMG-CoA reductase inhibitor.
  • High purity suitable for analytical and biochemical studies.
  • Available as a 25 mg pack for small-scale assays.
  • Molecular weight 453.61 g·mol⁻1 and formula C25H43NO6.
  • Used to study cholesterol biosynthesis, ROS modulation, and transporter expression.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.