Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Simvastatin acid ammonium | 139893-43-9 | 99.7% | 5 MG
SDP

Catalog No. 5000233325
Encompass_Preferred

Simvastatin acid ammonium is the ammonium salt form of simvastatin hydroxy acid, a potent HMG-CoA reductase inhibitor supplied as a high-purity research reagent for biochemical and pharmacological studies.

  • High purity (99.7%).
  • Chemical formula C25H43NO6.
  • Molecular weight 453.61 g/mol.
  • Provided as a solid powder suitable for dissolution or formulation.
  • Available in small research pack sizes for laboratory use.

Catalog No. 50-002-33325 Supplier Medchemexpress LLC Supplier No. HY119695A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Simvastatin acid ammonium is the ammonium salt form of simvastatin hydroxy acid, a potent HMG-CoA reductase inhibitor supplied as a high-purity research reagent for biochemical and pharmacological studies.

  • High purity (99.7%).
  • Chemical formula C25H43NO6.
  • Molecular weight 453.61 g/mol.
  • Provided as a solid powder suitable for dissolution or formulation.
  • Available in small research pack sizes for laboratory use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.