Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Simvastatin acid ammonium | 139893-43-9 | 99.7% | 50 MG
SDP

Catalog No. 5000233320
Encompass_Preferred

Simvastatin acid ammonium is the ammonium salt of simvastatin hydroxy acid, a potent HMG-CoA reductase inhibitor used as a research reagent to study cholesterol biosynthesis and related pathways. Supplied as a high-purity solid for laboratory research use; not for human consumption.

  • Ammonium salt of simvastatin hydroxy acid used for research purposes.
  • CAS 139893-43-9.
  • Molecular formula C25H43NO6, molecular weight 453.61 g·mol⁻¹.
  • High purity: 99.7%.
  • Supplied as a 50 MG quantity.
  • Activity: HMG-CoA reductase inhibitor relevant to cholesterol metabolism studies.

Catalog No. 50-002-33320 Supplier Medchemexpress LLC Supplier No. HY119695A50MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Simvastatin acid ammonium is the ammonium salt of simvastatin hydroxy acid, a potent HMG-CoA reductase inhibitor used as a research reagent to study cholesterol biosynthesis and related pathways. Supplied as a high-purity solid for laboratory research use; not for human consumption.

  • Ammonium salt of simvastatin hydroxy acid used for research purposes.
  • CAS 139893-43-9.
  • Molecular formula C25H43NO6, molecular weight 453.61 g·mol⁻¹.
  • High purity: 99.7%.
  • Supplied as a 50 MG quantity.
  • Activity: HMG-CoA reductase inhibitor relevant to cholesterol metabolism studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.