Learn More
Medchemexpress LLC Simvastatin-d3 | 1002347-61-6 | 98.6% | 421.58 | 1 MG
SDP
Catalog No. 5000386768
Encompass_Preferred
Simvastatin-d3 is the deuterium-labeled Simvastatin. Simvastatin (MK 733) is a competitive inhibitor of HMG-CoA reductase with a Ki of 0.2 nM. It is for research use only and not sold to patients.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process.
  • Deuteration has the potential to affect the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-003-86768 Supplier Medchemexpress LLC Supplier No. HY17502S11MG
Simvastatin-d3 is the deuterium-labeled Simvastatin. Simvastatin (MK 733) is a competitive inhibitor of HMG-CoA reductase with a Ki of 0.2 nM. It is for research use only and not sold to patients.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process.
  • Deuteration has the potential to affect the pharmacokinetic and metabolic profiles of drugs.