Learn More
Medchemexpress LLC Simvastatin-d3 | 1002347-61-6 | 98.6% | 421.58 | 5 MG
SDP
Catalog No. 5000386769
Encompass_Preferred
Simvastatin-d3 is a deuterium-labeled version of Simvastatin, a competitive inhibitor of HMG-CoA reductase with a Ki of 0.2 nM. It is designed for research use only and is not intended for patient use. This compound can serve as a tracer and an internal standard for quantitative analysis by techniques such as NMR, GC-MS, or LC-MS.
  • Deuterium labeled Simvastatin
  • Competitive inhibitor of HMG-CoA reductase
  • Used as a tracer in drug development
  • Used as an internal standard for quantitative analysis
  • Impacts pharmacokinetic and metabolic profiles of drugs
  • Suitable for NMR, GC-MS, or LC-MS applications
Catalog No. 50-003-86769 Supplier Medchemexpress LLC Supplier No. HY17502S15MG
Simvastatin-d3 is a deuterium-labeled version of Simvastatin, a competitive inhibitor of HMG-CoA reductase with a Ki of 0.2 nM. It is designed for research use only and is not intended for patient use. This compound can serve as a tracer and an internal standard for quantitative analysis by techniques such as NMR, GC-MS, or LC-MS.
  • Deuterium labeled Simvastatin
  • Competitive inhibitor of HMG-CoA reductase
  • Used as a tracer in drug development
  • Used as an internal standard for quantitative analysis
  • Impacts pharmacokinetic and metabolic profiles of drugs
  • Suitable for NMR, GC-MS, or LC-MS applications