Learn More
Medchemexpress LLC Simvastatin-d3 | 1002347-61-6 | MFCD09832920 | 98.6% | 421.58 g/mol | C25H35D3O5 | 10 MG
SDP
Catalog No. 5000298533
Encompass_Preferred
Simvastatin-d3 is the deuterium-labeled form of simvastatin used as a stable isotope tracer and internal standard for quantitative analysis by NMR, GC-MS, and LC-MS. It is supplied as a white to off-white solid with high purity and defined storage stability for powder and solutions, making it suitable for analytical and research applications.
  • Deuterium-labeled internal standard for quantitative assays.
  • Suitable for NMR, GC-MS, and LC-MS applications.
  • High purity (98.6%) for accurate quantitation.
  • White to off-white solid form.
  • Defined storage stability for powder and in solvent.
Catalog No. 50-002-98533 Supplier Medchemexpress LLC Supplier No. HY17502S110MG
Simvastatin-d3 is the deuterium-labeled form of simvastatin used as a stable isotope tracer and internal standard for quantitative analysis by NMR, GC-MS, and LC-MS. It is supplied as a white to off-white solid with high purity and defined storage stability for powder and solutions, making it suitable for analytical and research applications.
  • Deuterium-labeled internal standard for quantitative assays.
  • Suitable for NMR, GC-MS, and LC-MS applications.
  • High purity (98.6%) for accurate quantitation.
  • White to off-white solid form.
  • Defined storage stability for powder and in solvent.