Learn More
Cayman Chemical Simvastatin-d6, Purity: ≥99% deuterated forms (d1-d6), CAS: 1002347-71-8, 10 mg
SDP
Catalog No. 502986351
Encompass_Preferred
An internal standard for the quantification of simvastatin by GC- or LC-MS
Catalog No. 50-298-6351 Supplier Cayman Chemical Supplier No. 2218110 MG
An internal standard for the quantification of simvastatin by GC- or LC-MS