Learn More
Medchemexpress LLC Simvastatin-d6 hydroxy acid, ammonium salt | 139893-43-9 | >95.0% | 459.65 | C25H37D6NO6 | 1 MG

Supplier: Medchemexpress LLC HYW6543411MG
Simvastatin-d6 hydroxy acid ammonium is a deuterium-labeled simvastatin hydroxy-acid (ammonium salt) supplied for research use as an analytical internal standard and tracer. It supports quantitative analysis in NMR, GC-MS, and LC-MS workflows and is provided in small reference quantities for laboratory analytical applications.
- Deuterium-labeled hydroxy acid ammonium salt for use as an internal standard.
- Suitable as a tracer and internal standard for quantitative NMR, GC-MS, and LC-MS analyses.
- Available in small reference quantities for analytical workflows.
- Molecular formula C25H37D6NO6; molecular weight 459.65.
- Purity >95.0% (HPLC) typical for reference standards; consult certificate of analysis for exact value.
- Store under recommended conditions indicated on the certificate of analysis.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.