Learn More
Cayman Chemical SImvastatIn Hydroxy ACDam 10mg
SDP
Catalog No. 502911446
Encompass_Preferred
An inhibitor of HMG-CoA reductase (Ki = 0.12 nM); an active metabolite of simvastatin
Catalog No. 50-291-1446 Supplier Cayman Chemical Supplier No. 3488310 MG
An inhibitor of HMG-CoA reductase (Ki = 0.12 nM); an active metabolite of simvastatin