Learn More
Cayman Chemical SImvastatIn Hydroxy ACDam 50mg
SDP
Supplier:  Cayman Chemical 3488350 MG
Encompass_Preferred
An inhibitor of HMG-CoA reductase (Ki = 0.12 nM); an active metabolite of simvastatin
Catalog No. 50-291-1325