Learn More
AdipoGen Simvastatin sodium salt
SDP
Catalog No. 502562488
Encompass_Preferred
Chemical. CAS 101314-97-0. Formula C25H39O6 . Na. MW 458.6. Synthetic analog of Lovastatin. Water-soluble carboxylate form of Simvastatin. Potent competitive HMG-CoA reductase inhibitor. Anti-hypercholesterolemic agent. Cholesterol/isoprenoid biosynthesis inhibitor. Blocks the production of mevalonate, a critical compound in the production of cholesterol and isoprenoids. Inhibits the isoprenylation of Ras and Rho family GTPases. Smooth muscle cell proliferation inhibitor. Anti-adhesive, immunomodulatory and anti-inflammatory compound. Proteasome modulator. Stimulates bone formation. Apoptosis inducer. Anticancer compound. Increases cellular resistance to anticancer agents such as doxorubicin and etoposides.
Catalog No. 50-256-2488 Supplier AdipoGen Supplier No. AGCN20539M025
Chemical. CAS 101314-97-0. Formula C25H39O6 . Na. MW 458.6. Synthetic analog of Lovastatin. Water-soluble carboxylate form of Simvastatin. Potent competitive HMG-CoA reductase inhibitor. Anti-hypercholesterolemic agent. Cholesterol/isoprenoid biosynthesis inhibitor. Blocks the production of mevalonate, a critical compound in the production of cholesterol and isoprenoids. Inhibits the isoprenylation of Ras and Rho family GTPases. Smooth muscle cell proliferation inhibitor. Anti-adhesive, immunomodulatory and anti-inflammatory compound. Proteasome modulator. Stimulates bone formation. Apoptosis inducer. Anticancer compound. Increases cellular resistance to anticancer agents such as doxorubicin and etoposides.