Learn More
Cayman Chemical Simvastatin (sodium salt), Purity: ≥98%, CAS: 101314-97-0, 5 mg
SDP
Catalog No. 502889201
Encompass_Preferred
An HMG-CoA reductase inhibitor (Ki = 0.12 nM); reduces plasma cholesterol levels in rats and dogs at 1.2 and 8 mg/kg, respectively; suppresses TNF-induced NF-κB activation (IC50 = ~13 µM) and potentiates apoptosis in human myeloid leukemia cells; inhibits GPX4, increases MDA levels, and induces ferroptosis in MDA-MB-231 and MCF-7 breast cancer cells
Catalog No. 50-288-9201 Supplier Cayman Chemical Supplier No. 100103455 MG
An HMG-CoA reductase inhibitor (Ki = 0.12 nM); reduces plasma cholesterol levels in rats and dogs at 1.2 and 8 mg/kg, respectively; suppresses TNF-induced NF-κB activation (IC50 = ~13 µM) and potentiates apoptosis in human myeloid leukemia cells; inhibits GPX4, increases MDA levels, and induces ferroptosis in MDA-MB-231 and MCF-7 breast cancer cells