Learn More
Enzo Life Sciences Simvastatin. sodium salt (5mg). CAS: 101314-97-0
SDP
Catalog No. 502013443
Encompass_Preferred
Simvastatin is marketed under the trade name Zocor™ and is often prescribed in combination with ezetimibe (Zetia) to treat dyslipidemia. This drug combination is known commercially as Vytorin or Inegy. Simvastatin also suppresses TNF-induced NF-κB activation, which potentiates apoptosis in human myeloid leukemia cells and thus, may be useful in treating cancer. Alternative name: (betaR,deltaR,​1S,​2S,​6R,​8S,​8aR)-​8-​(2,​2-​dimethyl-​1-​oxobutoxy)-​1,​2,​6,​7,​8,​8a-​hexahydro-​beta,delta-​dihydroxy-​2,​6-​dimethyl-​1-​naphthaleneheptanoic acid .​ monosodium salt. Purity: ≥97% (HPLC). Solubility: Soluble in water. Long Term Storage: -80°C.
Catalog No. 50-201-3443 Supplier Enzo Life Sciences Supplier No. ENZCHM1420005
Simvastatin is marketed under the trade name Zocor™ and is often prescribed in combination with ezetimibe (Zetia) to treat dyslipidemia. This drug combination is known commercially as Vytorin or Inegy. Simvastatin also suppresses TNF-induced NF-κB activation, which potentiates apoptosis in human myeloid leukemia cells and thus, may be useful in treating cancer. Alternative name: (betaR,deltaR,​1S,​2S,​6R,​8S,​8aR)-​8-​(2,​2-​dimethyl-​1-​oxobutoxy)-​1,​2,​6,​7,​8,​8a-​hexahydro-​beta,delta-​dihydroxy-​2,​6-​dimethyl-​1-​naphthaleneheptanoic acid .​ monosodium salt. Purity: ≥97% (HPLC). Solubility: Soluble in water. Long Term Storage: -80°C.