Learn More
Apexbio Technology LLC Simvastatin (Zocor), 79902-63-9, MFCD00072007, 50 mg
SDP
Supplier:  Apexbio Technology LLC A852250
Encompass_Preferred
MF: C25H38O5, MW: 418.6, Solubility: 20.95mg/mL in DMSO. Simvastatin (SIM), a lactone, is white, crystalline, nonhygroscopic and powdery, practically insoluble in water (30 mcg/ml), and 0.1 (N) HCl (60 mcg/ml).
Catalog No. 50-101-0528