Learn More
AdipoGen SIN-1 chloride (10 mg)
SDP
Supplier:  AdipoGen AGCR10027M010
Encompass_Preferred
Chemical. CAS: 16142-27-1. Formula: C6H11ClN4O2. MW: 206.6. Nitric oxide (NO) donor. Generates both superoxide anion and nitric oxide that spontaneously form peroxynitrite. Platelet aggregation inhibitor. Guanylyl cyclase activator. Cytotoxic. Positive ionotropic. Vasodilator.
Catalog No. 50-205-3777