Learn More
AdipoGen SIN-1 chloride,Chemical. CAS: 16142-27-1
SDP
Supplier:  AdipoGen AGCR10027M050
Encompass_Preferred
SIN-1 chloride, AdipoGen Life Sciences. Chemical. CAS: 16142-27-1. Formula: C6H11ClN4O2. MW: 206.6. Nitric oxide (NO) donor. Generates both superoxide anion and nitric oxide that spontaneously form peroxynitrite. Platelet aggregation inhibitor. Guanylyl cyclase activator. Cytotoxic. Positive ionotropic. Vasodilator.
Catalog No. 50-464-585