Learn More
Enzo Life Sciences SIN-1A/gammaCD Complex (25mg). CAS: 26687-79-6 (SIN-1A)
SDP
Supplier:  Enzo Life Sciences ALX400009M025
Encompass_Preferred
Physiologically active nitric oxide (NO) releasing agent. Releasing one equivalent of NO the half life time at room temperature is ~40 minutes mostly independent from the pH. Alternative name: 3-Morpholino-N-nitroso-aminoacetonitrile . gammacyclodextrin complex. Formulation: The complex contains 10 ± 2% of SIN-1A. SIN-1 content: not detectable. SIN-1C (N-morpholinoiminoacetonitrile) content: 0.5 ± 0.2%. Solubility: Well soluble in distilled water, isotonic saline, phosphate buffer or HCl solution, pH 1.4. Long Term Storage: +4°C.
Catalog No. 50-200-9068