Learn More
Cayman Chemical SIN-1chlorIde 100mg
SDP
Catalog No. 502896611
Encompass_Preferred
Active metabolite of molsidomine that acts as a potent vasorelaxant and inhibitor of platelet aggregation; produces both NO and superoxide to generate peroxynitrite under physiological conditions
Catalog No. 50-289-6611 Supplier Cayman Chemical Supplier No. 82220100 MG
Active metabolite of molsidomine that acts as a potent vasorelaxant and inhibitor of platelet aggregation; produces both NO and superoxide to generate peroxynitrite under physiological conditions