Learn More
Cayman Chemical SIN-1chlorIde 10mg
SDP
Catalog No. 502896680
Encompass_Preferred
Active metabolite of molsidomine that acts as a potent vasorelaxant and inhibitor of platelet aggregation; produces both NO and superoxide to generate peroxynitrite under physiological conditions
Catalog No. 50-289-6680 Supplier Cayman Chemical Supplier No. 8222010 MG
Active metabolite of molsidomine that acts as a potent vasorelaxant and inhibitor of platelet aggregation; produces both NO and superoxide to generate peroxynitrite under physiological conditions