Learn More
Cayman Chemical SIN-1 (chloride), Purity: ≥99%, CAS: 16142-27-1, 50 mg
SDP
Catalog No. 502896603
Encompass_Preferred
Active metabolite of molsidomine that acts as a potent vasorelaxant and inhibitor of platelet aggregation; produces both NO and superoxide to generate peroxynitrite under physiological conditions
Catalog No. 50-289-6603 Supplier Cayman Chemical Supplier No. 8222050 MG
Active metabolite of molsidomine that acts as a potent vasorelaxant and inhibitor of platelet aggregation; produces both NO and superoxide to generate peroxynitrite under physiological conditions