Learn More
Aviva Systems Biology Corp SIN3A Antibody - N-terminal region (ARP32483_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32483_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SIN3A. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human SIN3A. Alternate names: DKFZP434K2235, FLJ90319, KIAA0700ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-54617